missing translation for 'onlineSavingsMsg'
Learn More

METTLER TOLEDO™ InLab™ Reach pH Electrodes

A pH electrode with an integrated temperature sensor featuring an extended shaft for measuring a wide range of samples in large vessels.

Specifications

Junction Ceramic
Material Glass
pH Range 0 to 14
Type Combination Electrode
For Use With (Application) Accurate measurement in a wide range of sample types
View More Specs

Products 6
Catalog Number Mfr. No. Connector Types Includes Length (Metric) Stem Price Quantity & Availability  
Catalog Number Mfr. No. Connector Types Includes Length (Metric) Stem Price Quantity & Availability  
01911473
View Documents Promotion Details
METTLER TOLEDO™
51343062
Multi-Pin Temperature Probe (Pt-1000) 425 mm N/A
Only null left
 
01911472
View Documents Promotion Details
METTLER TOLEDO™
51343061
Multi-Pin Temperature Probe (NTC 30 kΩ) 425 mm N/A
Only null left
 
01914302
View Documents Promotion Details
METTLER TOLEDO™
30244733
S7 Electrode only 225 mm N/A
Only null left
 
01914303
View Documents Promotion Details
METTLER TOLEDO™
30248120
S7 Electrode only 425 mm N/A
Only null left
 
01914304
View Documents Promotion Details
METTLER TOLEDO™
30248826
Multi-Pin Temperature Probe (NTC 30 kΩ) 225 mm N/A
Only null left
 
01914305
View Documents Promotion Details
METTLER TOLEDO™
30248828
Multi-Pin Temperature Probe (Pt-1000) 225 mm N/A
Only null left
 
Description

Description

  • For use with METTLER TOLEDO SevenExcellence and SevenCompact meters as well as other brands. Connection cables may be required.
  • Capture electrode calibration and results with LabX software, designed for advanced data management and 21CFR part 11 regulatory compliance.
  • Compatible with InMotion Autosampler to automate measurement and cleaning of up to 303 samples.

Featured Applications

  • Pilot reactor
  • Tank and Barrel
  • Large samples
  • Deep vessels

Accessories & Care

  • Ceramic junction
  • HA glass membrane
  • Refillable 3M KCl (51350072): Keep probe filled with electrolyte for accurate results
  • Store in 3M KCl (51350072) or InLab Storage Solution (30111142)
  • Recommended for use with METTLER TOLEDO buffers (Sachets: 51302080, Bottles: 63056116)
Specifications

Specifications

Ceramic
0 to 14
Accurate measurement in a wide range of sample types
0°C to 100°C
Glass
Combination Electrode
HA Glass
Videos
SDS
Documents

Documents