missing translation for 'onlineSavingsMsg'
Learn More
Learn More
METTLER TOLEDO™ InLab™ Reach pH Electrodes
A pH electrode with an integrated temperature sensor featuring an extended shaft for measuring a wide range of samples in large vessels.
Supplier: METTLER TOLEDO™ 51343062
Description
- For use with METTLER TOLEDO SevenExcellence and SevenCompact meters as well as other brands. Connection cables may be required.
- Capture electrode calibration and results with LabX software, designed for advanced data management and 21CFR part 11 regulatory compliance.
- Compatible with InMotion Autosampler to automate measurement and cleaning of up to 303 samples.
Featured Applications
- Pilot reactor
- Tank and Barrel
- Large samples
- Deep vessels
Accessories & Care
- Ceramic junction
- HA glass membrane
- Refillable 3M KCl (51350072): Keep probe filled with electrolyte for accurate results
- Store in 3M KCl (51350072) or InLab Storage Solution (30111142)
- Recommended for use with METTLER TOLEDO buffers (Sachets: 51302080, Bottles: 63056116)
Chemical Identifiers
| Ceramic | |
| 0 to 14 | |
| Accurate measurement in a wide range of sample types | |
| 0°C to 100°C |
| Glass | |
| Combination Electrode | |
| HA Glass |
Specifications
| pH electrode InLab Reach Pt1000-425 | |
| Ceramic | |
| 0 to 14 | |
| Combination Electrode | |
| Accurate measurement in a wide range of sample types | |
| Temperature Probe (Pt-1000) | |
| HA Glass | |
| PT1000 | |
| 3mol/L KCl | |
| 0°C to 100°C | |
| 400 mm | |
| 0.47 in. |
| Multi-Pin | |
| Glass | |
| 0°C to 100°C | |
| 3mol/L KCl | |
| Multi-Pin to BNC (30281896) | |
| ARGENTHAL with Ag+ trap | |
| InLab Reach Pt1000-425 | |
| <20 sec. | |
| 32°F to 212°F | |
| 15.7 in. | |
| 425 mm | |
| 12 mm |