missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Inkjet Vinyl Label and Overlaminate Kit
Take your inkjet printing further and label faster with these ideally sized label and overlaminate kits.
$513.79 - $751.61
Specifications
| Autoclavable | Not Autoclavable |
|---|---|
| Adhesive Type | Permanent Acrylic |
| Coating | None |
| Color | White |
| Finish | Matte |
| Catalog Number | Mfr. No. | Dimensions (L x W) | For Use With (Equipment) | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Dimensions (L x W) | For Use With (Equipment) | Price | Quantity & Availability | ||||
|
17377057
|
Brady™
J202602595KT |
3.35 x 2.17 in. | J2000 Printer |
Each for $513.79
|
|
||||
|
17377028
|
Brady™
J502602595KT |
3.35 x 2.17 in. | J4000, J5000 Printer |
Each for $520.21
|
|
||||
|
17377055
|
Brady™
J202612595KT |
4.33 x 2.95 in. | J2000 Printer |
Each for $604.72
|
|
||||
|
17377026
|
Brady™
J502612595KT |
4.33 x 2.95 in. | J4000, J5000 Printer |
Each for $609.63
|
|
||||
|
17377053
|
Brady™
J202622595KT |
6.1 x 4.13 in. | J2000 Printer |
Each for $743.65
|
|
||||
|
17377024
|
Brady™
J502622595KT |
6.1 x 4.13 in. | J4000, J5000 Printer |
Each for $751.61
|
|
||||
|
17377020
|
Brady™
J502672595KT |
8.3 x 6.1 in. | J4000, J5000 Printer |
Each for $667.43
|
|
||||
|
17377022
|
Brady™
J502652595KT |
10 x 7 in. | J4000, J5000 Printer |
Each for $539.64
|
|
||||
Description
- Inkjet-printable label is outfitted with a specially formulated topcoat that invigorates colors and label appearance
- Label topcoat also protects against smudges, smears and splotches
- Acrylic overlaminate provides up to 10 years of outdoor durability through improved UV and abrasion resistance
- Overlaminate is oversized by a quarter-inch for easier application and complete coverage
- An ink cartridge is required to print label material
Specifications
| Not Autoclavable | |
| None | |
| Matte | |
| Vinyl | |
| Non-sterile | |
| 2-roll Kit | |
| Rectangle | |
| -40°C to +60°C | |
| 0.1372 mm |
| Permanent Acrylic | |
| White | |
| Die-cut | |
| Ink Jet | |
| Facility Identification, General Identification, Hazard Communication Identification, Lean and 5S Labeling, Lockout Tagout, Pipe and Valve Marking, Warehouse Marking | |
| -40°C | |
| -40°F to +140°F | |
| 0.0054 in. |