missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Inkjet Vinyl Label and Overlaminate Kit

Catalog No. 17377057
Change view
Click to view available options
Dimensions (L x W):
3.35 x 2.17 in.
4.33 x 2.95 in.
6.1 x 4.13 in.
8.3 x 6.1 in.
10 x 7 in.
For Use With (Equipment):
J2000 Printer
J4000, J5000 Printer
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W) For Use With (Equipment)
17377057 3.35 x 2.17 in. J2000 Printer
17377028 3.35 x 2.17 in. J4000, J5000 Printer
17377055 4.33 x 2.95 in. J2000 Printer
17377026 4.33 x 2.95 in. J4000, J5000 Printer
17377053 6.1 x 4.13 in. J2000 Printer
17377024 6.1 x 4.13 in. J4000, J5000 Printer
17377020 8.3 x 6.1 in. J4000, J5000 Printer
17377022 10 x 7 in. J4000, J5000 Printer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 17377057 Supplier Brady™ Supplier No. J202602595KT

Take your inkjet printing further and label faster with these ideally sized label and overlaminate kits.

  • Inkjet-printable label is outfitted with a specially formulated topcoat that invigorates colors and label appearance
  • Label topcoat also protects against smudges, smears and splotches
  • Acrylic overlaminate provides up to 10 years of outdoor durability through improved UV and abrasion resistance
  • Overlaminate is oversized by a quarter-inch for easier application and complete coverage
  • An ink cartridge is required to print label material

Specifications

Autoclavable Not Autoclavable
Adhesive Type Permanent Acrylic
Coating None
Color White
Finish Matte
Format Die-cut
Material Vinyl
Printable Sides Single-side Printable
Print Technology Ink Jet
Product Type Label
Sterility Non-sterile
For Use With (Application) Facility Identification, General Identification, Hazard Communication Identification, Lean and 5S Labeling, Lockout Tagout, Pipe and Valve Marking, Warehouse Marking
For Use With (Equipment) J2000 Printer
Includes 2-roll Kit
Min. Operating Temperature (Metric) -40°C
Quantity 340 Labels
Shape Rectangle
Size (Metric) 85.09 x 55.12 mm
Temperature Range (English) -40°F to +140°F
Temperature Range (Metric) -40°C to +60°C
Thickness (English) 0.0054 in.
Thickness (Metric) 0.1372 mm
Dimensions (L x W) 3.35 x 2.17 in.
Length (English) 3.35 in.
Length (Metric) 85.09 mm
Width (English) 2.17 in.
Width (Metric) 55.12 mm
Show More Show Less