missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Inhibitor Removal Resin

Catalog No. AA4248914
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4248914 25 g
AA4248922 100 g
AA4248936 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4248914 Supplier Thermo Scientific Chemicals Supplier No. 04248914

CAS: 9003-70-7 | C24H27NO5 | 409.48 g/mol

Inhibitor removal resin is used for the removal of phenolic inhibitors: hydroquinone, hydroquinone monomethyl ether, tert-butylcatechol. It is also take part in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 9003-70-7

Specifications

Product Type Inhibitor Removal Resin
Odor Odorless
Molecular Formula C24H27NO5
MDL Number 9003-70-7
Form Crystalline Solid
For Use With (Application) For the removal of phenolic inhibitors: hydroquinone, hydroquinone monomethyl ether, tert-butylcatechol
InChI Key BCIPGSZQUDLGSY-UHFFFAOYNA-N
SMILES CC(C)(C)OC(=O)CCC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C=O
IUPAC Name tert-butyl 4-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-5-oxopentanoate
Molecular Weight (g/mol) 409.48
Quantity 25 g
Show More Show Less
Recommended Storage Ambient temperatures

RUO – Research Use Only