missing translation for 'onlineSavingsMsg'
Learn More

ING1 Monoclonal Antibody (K06325_6F4), Invitrogen™

Mouse Monoclonal Antibody

$1601.06 - $2821.92

Specifications

Antigen ING1
Clone K06325_6F4
Concentration 1 mg/mL
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
MA556060
View Documents
Invitrogen™
MA556060
200 μL
Each for $2,821.92
Only null left
 
MA555680
View Documents
Invitrogen™
MA555680
50 μL
Each for $1,601.06
Only null left
 
Description

Description

Sequence of this protein is as follows: AAAQADKPNS KRSRRQRNNE NRENASSNHD HDDGASGTPK EKKAKTSKKK KRSKAKAERE ASPADLPIDP NEPTYCLCNQ VSYGEMIGCD NDECPIEWFH FSCVGLNHKP KGKWYCPKCR GENEKTMDKA

ING1 Monoclonal antibody specifically detects ING1 in Human, Mouse samples. It is validated for Immunoprecipitation, Western Blot
Specifications

Specifications

ING1
1 mg/mL
Monoclonal
Liquid
RUO
PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
2610028J21Rik; AA407184; AI875420; growth inhibitor ING1; growth inhibitory protein ING1; ING1; inhibitor of growth family member 1; inhibitor of growth family, member 1; inhibitor of growth protein 1; mING1h; p24ING1c; p33; p33ING; p33ING1; p33ING1b; p33ING1c; p37Ing1b; p47; p47ING1a; tumor suppressor ING1
Recombinant human ING1 (immunogen range is 281-410).
Primary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
ING1
K06325_6F4
Immunoprecipitation, Western Blot
Unconjugated
Mouse
Human, Mouse
Q9QXV3, Q9UK53
26356, 3621
IgG2b
Protein A
Antibody
Videos
SDS
Documents

Documents