missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ING1 Monoclonal Antibody (K06325_6F4), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA555680
Description
Sequence of this protein is as follows: AAAQADKPNS KRSRRQRNNE NRENASSNHD HDDGASGTPK EKKAKTSKKK KRSKAKAERE ASPADLPIDP NEPTYCLCNQ VSYGEMIGCD NDECPIEWFH FSCVGLNHKP KGKWYCPKCR GENEKTMDKA
This gene encodes a tumor suppressor protein that can induce cell growth arrest and apoptosis. The encoded protein is a nuclear protein that physically interacts with the tumor suppressor protein TP53 and is a component of the p53 signaling pathway. Reduced expression and rearrangement of this gene have been detected in various cancers. Multiple alternatively spliced transcript variants encoding distinct isoforms have been reported.Specifications
| ING1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| Q9QXV3, Q9UK53 | |
| ING1 | |
| Recombinant human ING1 (immunogen range is 281-410). | |
| 50 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG2b |
| Immunoprecipitation, Western Blot | |
| K06325_6F4 | |
| Unconjugated | |
| ING1 | |
| 2610028J21Rik; AA407184; AI875420; growth inhibitor ING1; growth inhibitory protein ING1; ING1; inhibitor of growth family member 1; inhibitor of growth family, member 1; inhibitor of growth protein 1; mING1h; p24ING1c; p33; p33ING; p33ING1; p33ING1b; p33ING1c; p37Ing1b; p47; p47ING1a; tumor suppressor ING1 | |
| Mouse | |
| Protein A | |
| RUO | |
| 26356, 3621 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |