missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| Indoline | |
| Yellow | |
| C8H9N | |
| 25 mL | |
| LPAGFVYQRIESJQ-UHFFFAOYSA-N | |
| 2,3-dihydro-1H-indole | |
| 10328 | |
| 119.17 | |
| Liquid |
| 496-15-1 | |
| 231°C | |
| MFCD00005705 | |
| indoline, 2,3-dihydroindole, 1-azaindan, dihydroindole, 1h-indole, 2,3-dihydro, azaindane, benzopyrrolidine, aza-indane, indoline, 19, pubchem7502 | |
| C1CNC2=CC=CC=C21 | |
| 119.167 | |
| CHEBI:43295 | |
| ≥98.0% (GC,T) |
Chemical Identifiers
| 496-15-1 | |
| 119.167 | |
| LPAGFVYQRIESJQ-UHFFFAOYSA-N | |
| 10328 | |
| 2,3-dihydro-1H-indole |
| C8H9N | |
| MFCD00005705 | |
| indoline, 2,3-dihydroindole, 1-azaindan, dihydroindole, 1h-indole, 2,3-dihydro, azaindane, benzopyrrolidine, aza-indane, indoline, 19, pubchem7502 | |
| CHEBI:43295 | |
| C1CNC2=CC=CC=C21 |
Safety and Handling
EINECSNumber : (5)-3364
RTECSNumber : NL6906300
TSCA : Yes