missing translation for 'onlineSavingsMsg'
Learn More

Indoline 98.0+%, TCI America™

Catalog No. p-7131171
Change view
Click to view available options
Quantity:
25 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I003325ML 25 mL
I0033250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I003325ML Supplier TCI America Supplier No. I003325ML
Only null left

Chemical Identifiers

CAS 496-15-1
Molecular Formula C8H9N
Molecular Weight (g/mol) 119.167
MDL Number MFCD00005705
InChI Key LPAGFVYQRIESJQ-UHFFFAOYSA-N
Synonym indoline, 2,3-dihydroindole, 1-azaindan, dihydroindole, 1h-indole, 2,3-dihydro, azaindane, benzopyrrolidine, aza-indane, indoline, 19, pubchem7502
PubChem CID 10328
ChEBI CHEBI:43295
IUPAC Name 2,3-dihydro-1H-indole
SMILES C1CNC2=CC=CC=C21

Specifications

Color Yellow
Boiling Point 231°C
Quantity 25 mL
Formula Weight 119.17
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Indoline
EINECSNumber (5)-3364
RTECSNumber NL6906300
TSCA Yes