missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Indocyanine green

Catalog No. 361063938
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
361063938 10 mg
751050 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 361063938 Supplier Tocris Bioscience™ Supplier No. 7510/10

Near-infrared fluorescent dye

Indocyanine green is a near infrared (NIR) fluorescent dye operating in the 700-900 nm wavelength range. It also exhibits fluorescence emission in the shortwave infrared window (SWIR; 1,000-2,000 nm) which enables in vivo imaging, including intravital microscopy, non-invasive real-time imaging in blood and lymph vessels, and imaging of hepatobiliary clearance. Imaging of indocyanine green in SWIR shows increased sensitivity, contrast, and resolution of fine anatomical structures over NIR.

This product is supplied with a high degree of hydration and may contain residual inorganic salts, the amount of which are batch dependent. Please refer to the Certificate of Analysis to obtain the batch specific Net Product Content.

Specifications

Chemical Name or Material Sodium 4-(2-((1 E,3 E,5 E,7 Z)-7-(1,1-Dimethyl-3-(4-sulfonatobutyl)-1,3-dihydro-2 H-benzo[e]indol-2-ylidene)hepta-1,3,5-trien-1-yl)-1,1-dimethyl-1 H-benzo[e]indol-3-ium-3-yl)butane-1-sulfonate
CAS 3599-32-4
Quantity 10 mg
Target Fluorescent Dyes
Molecular Formula C43H47N2O6S2Na
Purity 95%