Learn More
Sartorius Individual Precise Calibration Weight
ASTM Class 1 Individual Precise Calibration Weight without Certificate.
Supplier:  Sartorius WI001MAC1N
 View more versions of this product
Catalog No. 14558761
Only null left
Description
Description
  • To prevent any damage to the surface of weights, we offer you a wide range of accessories for the proper handling, storage and cleaning of weights.
Specifications
Specifications
Calibration Mass
ASTM Class 1
1 mg
0.000002 Ib.
Stainless Steel
<0.01
Product Suggestions
Product Suggestions