Learn More
Sartorius Individual Precise Calibration Weight
Catalog No. p-7229307
Change view
Click to view available options
Weight (Metric):
1 mg
2 mg
5 mg
10 mg
20 mg
50 mg
100 mg
200 mg
500 mg
1 g
2 g
5 g
10 g
20 g
50 g
100 g
200 g
500 g
1 kg
2 kg
5 kg
10 kg
22 product options available for selection
Product selection table with 22 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Weight (Metric)
14558753 500 mg
14558754 200 mg
14558755 100 mg
14558756 50 mg
14558757 20 mg
14558758 10 mg
14558759 5 mg
14558760 2 mg
14558761 1 mg
14558763 10 kg
14558764 5 kg
14558765 2 kg
14558766 1 kg
14558767 500 g
14558768 200 g
14558769 100 g
14558770 50 g
14558771 20 g
14558772 10 g
14558773 5 g
14558774 2 g
14558775 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 22 options available.
22 options
Catalog No. 14558753 Supplier Sartorius Supplier No. WI500MAC1N
Only null left
ASTM Class 1 Individual Precise Calibration Weight without Certificate.
  • To prevent any damage to the surface of weights, we offer you a wide range of accessories for the proper handling, storage and cleaning of weights.
Specifications
Product Type Calibration Mass
Weight (English) 0.0011 Ib.
Classification ASTM Class 1
Material Stainless Steel
Weight (Metric) 500 mg
Tolerance <0.01