missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Troemner™ Individual Analytical Precision Weights, Class 3 with Traceable Certificate
Low magnetic susceptibility, uniform consistency and high resistance to corrosion
Description
Designed for your most demanding weighing applications, Analytical Precision Weights meet or exceed specifications as defined in the ANSI/ASTM E617 standard.
- Used in extremely stable environments
- 1mg to 500mg weights are one-piece construction with no adjusting cavity with one end turned up for easy handling
- 1g to 5kg are two-piece construction with handling knob
- Surfaces are polished to a perfect, mirror like finish
- Appropriate for calibrating high-precision top loading balances with a readability as low as 0.01g to 0.001g
- Weights 1mg through 2kg are supplied in a high quality polycarbonate case
- Reference standards as well as calibrating Class I balances as specified in OIML R 76
- Calibrating balances with moderate precision, with a readability as low as 0.1g to 0.01g
Order Info
Protective cases that are supplied with weights can be ordered separately.
Specifications
| Metric Analytical Weight | |
| ASTM Class 3 | |
| 2 mg | |
| 2 mg | |
| Polycarbonate case |
| Traceable certificate, ISO9001 | |
| Aluminum | |
| 2.7g/cm3 at 20°C | |
| Flat with one side turned up | |
| Each |