missing translation for 'onlineSavingsMsg'
Learn More

Troemner™ Individual Analytical Precision Weights, Class 3 with Traceable Certificate

Catalog No. p-4515967
Change view
Click to view available options
Mass:
2 mg
5 mg
10 mg
20 mg
50 mg
100 mg
200 mg
300 mg
1 g
2 g
3 g
5 g
10 g
200 g
1 kg
2 kg
5 kg
Material:
Aluminum
Stainless Steel
Shape:
Cylindrical with Handling Knob
Flat with one side turned up
17 product options available for selection
Product selection table with 17 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Mass Material Shape
02225518 50 mg Stainless Steel Flat with one side turned up
02225529 2 mg Aluminum Flat with one side turned up
0221525DT 10 g Stainless Steel Cylindrical with Handling Knob
0221525AT 1 g Stainless Steel Cylindrical with Handling Knob
0221525CT 5 g Stainless Steel Cylindrical with Handling Knob
0221530DT 100 mg Stainless Steel Flat with one side turned up
02214084 300 mg Stainless Steel Flat with one side turned up
02225509 2 g Stainless Steel Cylindrical with Handling Knob
02214086 3 g Stainless Steel Cylindrical with Handling Knob
02214090 5 kg Stainless Steel Cylindrical with Handling Knob
02225514 200 mg Stainless Steel Flat with one side turned up
02214088 1 kg Stainless Steel Cylindrical with Handling Knob
02214089 2 kg Stainless Steel Cylindrical with Handling Knob
02225523 10 mg Stainless Steel Flat with one side turned up
02214087 200 g Stainless Steel Cylindrical with Handling Knob
02225521 20 mg Stainless Steel Flat with one side turned up
02225526 5 mg Stainless Steel Flat with one side turned up
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 17 options available.
17 options
Catalog No. 02225518
Only null left

Low magnetic susceptibility, uniform consistency and high resistance to corrosion

Designed for your most demanding weighing applications, Analytical Precision Weights meet or exceed specifications as defined in the ANSI/ASTM E617 standard.

  • Used in extremely stable environments
  • 1mg to 500mg weights are one-piece construction with no adjusting cavity with one end turned up for easy handling
  • 1g to 5kg are two-piece construction with handling knob
  • Surfaces are polished to a perfect, mirror like finish
  • Appropriate for calibrating high-precision top loading balances with a readability as low as 0.01g to 0.001g
  • Weights 1mg through 2kg are supplied in a high quality polycarbonate case
  • Reference standards as well as calibrating Class I balances as specified in OIML R 76
  • Calibrating balances with moderate precision, with a readability as low as 0.1g to 0.01g

Order Info

Protective cases that are supplied with weights can be ordered separately.

Specifications

Product Type Metric Analytical Weight
Certifications/Compliance Traceable certificate, ISO9001
Classification ASTM Class 3
Material Stainless Steel
Weight (Metric) 50 mg
Density 7.95g/cm3 at 20°C
Mass 50 mg
Shape Flat with one side turned up
Includes Polycarbonate case
Quantity Each