missing translation for 'onlineSavingsMsg'
Learn More

Troemner™ Individual Analytical Precision Weights, Class 3 with Traceable Certificate

Low magnetic susceptibility, uniform consistency and high resistance to corrosion

Supplier:  Troemner™ 80858261

 View more versions of this product

Catalog No. 0221525CT


Only null left
Explore available promotions

Description

Description

Designed for your most demanding weighing applications, Analytical Precision Weights meet or exceed specifications as defined in the ANSI/ASTM E617 standard.

  • Used in extremely stable environments
  • 1mg to 500mg weights are one-piece construction with no adjusting cavity with one end turned up for easy handling
  • 1g to 5kg are two-piece construction with handling knob
  • Surfaces are polished to a perfect, mirror like finish
  • Appropriate for calibrating high-precision top loading balances with a readability as low as 0.01g to 0.001g
  • Weights 1mg through 2kg are supplied in a high quality polycarbonate case
  • Reference standards as well as calibrating Class I balances as specified in OIML R 76
  • Calibrating balances with moderate precision, with a readability as low as 0.1g to 0.01g

Order Info

Protective cases that are supplied with weights can be ordered separately.

Specifications

Specifications

Metric Analytical Weight
ASTM Class 3
5 g
5 g
Polycarbonate case
1
Traceable certificate, ISO9001
Stainless Steel
7.95g/cm3 at 20°C
Cylindrical with Handling Knob
Each
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions