missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Indigo, synthetic

CAS: 482-89-3 | C16H10N2O2 | 262.268 g/mol

$75.99 - $264.85

Chemical Identifiers

CAS 482-89-3
Molecular Formula C16H10N2O2
Molecular Weight (g/mol) 262.268
MDL Number MFCD00005722
InChI Key COHYTHOBJLSHDF-BUHFOSPRSA-N
Synonym C.I. 73000, Vat Blue 1
PubChem CID 5318432
IUPAC Name (2E)-2-(3-oxo-1H-indol-2-ylidene)-1H-indol-3-one
SMILES C1=CC=C2C(=C1)C(=O)C(=C3C(=O)C4=CC=CC=C4N3)N2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
AC212130250
View Documents Promotion Details
Thermo Scientific Chemicals
212130250
25 g Glass bottle
Each for $75.99
Only null left
 
AC212131000
View Documents Promotion Details
Thermo Scientific Chemicals
212131000
100 g Glass bottle
Each for $264.85
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
Specifications

Chemical Identifiers

482-89-3
262.268
COHYTHOBJLSHDF-BUHFOSPRSA-N
5318432
C1=CC=C2C(=C1)C(=O)C(=C3C(=O)C4=CC=CC=C4N3)N2
C16H10N2O2
MFCD00005722
C.I. 73000, Vat Blue 1
(2E)-2-(3-oxo-1H-indol-2-ylidene)-1H-indol-3-one

Specifications

482-89-3
MFCD00005722
15, 4981
COHYTHOBJLSHDF-BUHFOSPRSA-N
Authentic
262.268
262.26
Blue to Violet
Indigo, synthetic
C16H10N2O2
24, 417
C.I. 73000, Vat Blue 1
C1=CC=C2C(=C1)C(=O)C(=C3C(=O)C4=CC=CC=C4N3)N2
(2E)-2-(3-oxo-1H-indol-2-ylidene)-1H-indol-3-one
5318432
Complies
Powder
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 207-586-9

RTECSNumber : DU2988400

TSCA : TSCA

SDS
Documents

Documents

RUO – Research Use Only