missing translation for 'onlineSavingsMsg'
Learn More

Indapamide

Catalog No. p-7042811
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6384603 1 g
AAJ6384606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6384603 Supplier Thermo Scientific Chemicals Supplier No. J6384603
Only null left

CAS: 26807-65-8 | C16H16ClN3O3S | 365.83 g/mol

Used as an antihypertensive. Diuretic

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Used as an antihypertensive
  • Diuretic

Chemical Identifiers

CAS 26807-65-8
Molecular Formula C16H16ClN3O3S
Molecular Weight (g/mol) 365.83
MDL Number MFCD00079375
InChI Key NDDAHWYSQHTHNT-UHFFFAOYNA-N
Synonym indapamide, noranat, veroxil, tertensif, arifon, fludex, lozol, indaflex, bajaten, indamol
PubChem CID 3702
ChEBI CHEBI:5893
IUPAC Name 4-chloro-N-(2-methyl-2,3-dihydro-1H-indol-1-yl)-3-sulfamoylbenzamide
SMILES CC1CC2=CC=CC=C2N1NC(=O)C1=CC=C(Cl)C(=C1)S(N)(=O)=O

Specifications

Melting Point 160°C to 162°C
Color White
Quantity 1 g
Merck Index 14,4935
Solubility Information Soluble in ethanol; Insoluble in water
Formula Weight 365.83
Physical Form Powder
Chemical Name or Material Indapamide
EINECSNumber 248-012-7
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only