missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Justrite™ In-flow Safety Cans
For use in storing reagents in approved containers when drawing solvents into HPLC machines
$1,648.41
Specifications
| Product Type | Safety Can |
|---|---|
| Certifications/Compliance | FM, NFPA, OSHA |
| Color | White |
| Material | Polyethylene |
| Includes | Self Closing Fill/Pour Spout |
| Catalog Number | Mfr. No. | Height (English) | Height (Metric) | Shape | Price | Quantity & Availability | |||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Height (English) | Height (Metric) | Shape | Price | Quantity & Availability | |||
|
19052391
|
Justrite™
12165 |
14.75 in. | 37.4 cm | Round |
Each for $1,648.41
|
|
|||
Description
- Made of high density polyethylene
- Equipped with a required self-closing fill/pour spout with stainless steel flame arrester
- Two quick-disconnect fittings: one polypropylene for venting, and one stainless steel to handle the solvent in-flow
- Stainless steel disconnect includes 1/8 in. OD (1/16 in.ID) and 1/16 in. OD (1/32 in. ID) compression fittings
Specifications
| Safety Can | |
| White | |
| Self Closing Fill/Pour Spout |
| FM, NFPA, OSHA | |
| Polyethylene |