missing translation for 'onlineSavingsMsg'
Learn More
Learn More
G-Biosciences Immobilized L-Tryptophan
Immobilized L-Tryptophan Resin Column is used for purification of amino acid binding proteins.
Supplier: G-Biosciences 7861377C
Description
- G-Biosciences offers pre-packed 5 mL Immobilized Amino Acids resin in a column format.
- Immobilized Amino Acids are prepared by coupling amino acid to preactivated 6% cross-linked agarose.
- The covalent coupling to agarose (resin) is achieved by cross-linking the amino or sulfhydryl group of the amino acid to the preactivated resin.
- Immobilized Amino Acids are used for purification of amino acid binding proteins.
Specifications
| 5 mL | |
| CaptureSelect | |
| Resin |
| Liquid Suspension | |
| 5 mL |