missing translation for 'onlineSavingsMsg'
Learn More
Learn More
G-Biosciences Immobilized L-Histidine
Description
- G-Biosciences offers pre-packed 5 mL Immobilized Amino Acids resin in a column format.
- Immobilized Amino Acids are prepared by coupling amino acid to preactivated 6% cross-linked agarose.
- The covalent coupling to agarose (resin) is achieved by cross-linking the amino or sulfhydryl group of the amino acid to the preactivated resin.
- Immobilized Amino Acids are used for purification of amino acid binding proteins.
Specifications
Specifications
| Form | Liquid Suspension |
| Product Line | CaptureSelect |
| Quantity | 5 mL |
| Type | Resin |
| Capacity (Metric) | 5 mL |