missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Immersion Oil for Microscopy

Eliminate light beam deflection to considerably increase the effectiveness of the lens. Immersion Oil for Microscopy is used with immersion objectives and is located between the specimen and the microscope lens. The refractive index is similar to that of glass.

$127.91 - $635.55

Specifications

Viscosity 100 to 120 mPas
Certifications/Compliance CE / IVD
Density 1.02g/cm3 (20°C)
Packaging Type Glass Bottle
Refractive Index 1.515 to 1.517 n 20/D
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
M1046990500
View Documents Promotion Details
MilliporeSigma™
1.04699.0500
500 mL
Each for $635.55
Only null left
 
M1046990100
View Documents Promotion Details
MilliporeSigma™
1.04699.0100
100 mL
Each for $127.91
Only null left
 
Description

Description

Note: Product comes in a glass bottle – not the plastic dropper bottle shown in the photo. 
Specifications

Specifications

100 to 120 mPas
1.02g/cm3 (20°C)
1.515 to 1.517 n 20/D
CE / IVD
Glass Bottle
Videos
SDS
Documents

Documents