Learn More
Fisherbrand™ Immersion Basket with Handle
Facilitates complete immersion within S-line Ultrasonic Cleaner baths. Fisherbrand™ Immersion Baskets with Handle are manufactured from stainless steel.
$214.81 - $1,011.54
| Catalog Number | Mfr. No. | For Use With (Equipment) | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | For Use With (Equipment) | Price | Quantity & Availability | |||||
|
2940705
|
Fisherbrand™
110 8916 |
Ultrasonic Cleaner, 2.75 L |
Each for $214.81
|
|
|||||
|
2940707
|
Fisherbrand™
111 3050 |
Ultrasonic Cleaner, 5.75 L |
Each for $342.20
|
|
|||||
|
2940708
|
Fisherbrand™
1004247 |
Ultrasonic Cleaner, 6.9 L |
Each for $653.27
|
|
|||||
|
2940711
|
Fisherbrand™
111 3390 |
Ultrasonic Cleaner, 12.75 L |
Each for $731.04
|
|
|||||
|
2940712
|
Fisherbrand™
111 3404 |
Ultrasonic Cleaner, 18 L |
Each for $824.38
|
|
|||||
|
2940713
|
Fisherbrand™
111 3072 |
Ultrasonic Cleaner, 28 L |
Each for $976.33
|
|
|||||
|
FB15080
|
Fisherbrand™
1004271 |
Ultrasonic Cleaner, 28 L |
Pack for $1,011.54
|
|
|||||
Description
Designed for the complete immersion of items and parts in Fisherbrand™ 112xxx Series Dual Frequency Ultrasonic Cleaners. Choose the Fisherbrand™ Stainless Steel Immersion Basket with Handles designed for the specific ultrasonic cleaner being used.
- Mesh bottom
- One handle on each end
- Functional and modern design
- Available for a variety of capacities