Learn More
Fisherbrand™ Immersion Basket with Handle
Facilitates complete immersion within S-line Ultrasonic Cleaner baths. Fisherbrand™ Immersion Baskets with Handle are manufactured from stainless steel.
Supplier: Fisherbrand™ 111 3050
Description
Designed for the complete immersion of items and parts in Fisherbrand™ 112xxx Series Dual Frequency Ultrasonic Cleaners. Choose the Fisherbrand™ Stainless Steel Immersion Basket with Handles designed for the specific ultrasonic cleaner being used.
- Designed for the Fisherbrand Dual Frequency Ultrasonic Cleaner models
- Fine mesh bottom keeps small parts secure
- Two handles for ease-of-use
- Clean and simple design fits perfectly
- Top-quality durable stainless steel
Specifications
| Basket | |
| Ultrasonic Cleaner, 5.75 L | |
| Stainless Steel |