missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Imidocarb Dipropionate 98.0+%, TCI America™
$68.97 - $274.02
Chemical Identifiers
| CAS | 55750-06-6 |
|---|---|
| Molecular Formula | C25H32N6O5 |
| Molecular Weight (g/mol) | 496.568 |
| MDL Number | MFCD00013161 |
| InChI Key | AFGQXWSHYUHHNV-UHFFFAOYSA-N |
| PubChem CID | 9983292 |
| IUPAC Name | 1,3-bis[3-(4,5-dihydro-1H-imidazol-2-yl)phenyl]urea;propanoic acid |
| SMILES | CCC(=O)O.CCC(=O)O.C1CN=C(N1)C2=CC(=CC=C2)NC(=O)NC3=CC=CC(=C3)C4=NCCN4 |
Chemical Identifiers
| 55750-06-6 | |
| 496.568 | |
| AFGQXWSHYUHHNV-UHFFFAOYSA-N | |
| 1,3-bis[3-(4,5-dihydro-1H-imidazol-2-yl)phenyl]urea;propanoic acid |
| C25H32N6O5 | |
| MFCD00013161 | |
| 9983292 | |
| CCC(=O)O.CCC(=O)O.C1CN=C(N1)C2=CC(=CC=C2)NC(=O)NC3=CC=CC(=C3)C4=NCCN4 |
Specifications
| 55750-06-6 | |
| White | |
| MFCD00013161 | |
| AFGQXWSHYUHHNV-UHFFFAOYSA-N | |
| 1,3-bis[3-(4,5-dihydro-1H-imidazol-2-yl)phenyl]urea;propanoic acid | |
| 9983292 | |
| ≥98.0% (HPLC,T) | |
| Imidocarb Dipropionate |
| 203°C | |
| C25H32N6O5 | |
| 1 g | |
| CCC(=O)O.CCC(=O)O.C1CN=C(N1)C2=CC(=CC=C2)NC(=O)NC3=CC=CC(=C3)C4=NCCN4 | |
| 496.568 | |
| 496.57 | |
| Crystalline Powder |
Safety and Handling
TSCA : No