missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Imidazole, Reagent (Glyoxaline), Reagents
Imidazole, Reagent (Glyoxaline), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Labchem Inc C2216600500A5
Specifications
| Imidazole, Reagent (Glyoxaline) | |
| 257°C | |
| White to Off-White | |
| 88°C to 90°C | |
| C3H4N2 | |
| 500 g | |
| 1,3-Diaza-2,4-cyclopentadiene, Glyoxaline | |
| N1C=CN=C1 | |
| 68.08 | |
| Solid |
| 288-32-4 | |
| 88°C to 90°C | |
| 257°C | |
| Poly Bottle | |
| MFCD00005183 | |
| 1.0303 | |
| RAXXELZNTBOGNW-UHFFFAOYSA-N | |
| 1H-imidazole | |
| Reagent |
Chemical Identifiers
| 288-32-4 | |
| 68.08 | |
| RAXXELZNTBOGNW-UHFFFAOYSA-N | |
| 1H-imidazole |
| C3H4N2 | |
| MFCD00005183 | |
| 1,3-Diaza-2,4-cyclopentadiene, Glyoxaline | |
| N1C=CN=C1 |