missing translation for 'onlineSavingsMsg'
Learn More

Imidazole, Reagent (Glyoxaline), Reagents

Catalog No. p-200021162
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337215 100 g
01337214 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01337215 Supplier Labchem Inc Supplier No. C2216600100C2
Only null left

Imidazole, Reagent (Glyoxaline), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 288-32-4
Molecular Formula C3H4N2
Molecular Weight (g/mol) 68.08
MDL Number MFCD00005183
InChI Key RAXXELZNTBOGNW-UHFFFAOYSA-N
Synonym 1,3-Diaza-2,4-cyclopentadiene, Glyoxaline
IUPAC Name 1H-imidazole
SMILES N1C=CN=C1

Specifications

Boiling Range 257°C
Melting Point 88°C to 90°C
Color White to Off-White
Boiling Point 257°C
Melting Range 88°C to 90°C
Packaging Glass Bottle
Quantity 100 g
Specific Gravity 1.0303
Grade Reagent
Physical Form Solid
Chemical Name or Material Imidazole, Reagent (Glyoxaline)
Show More Show Less