missing translation for 'onlineSavingsMsg'
Learn More

Imidazole, 0.5M buffer soln., pH 6.5, Thermo Scientific™

Catalog No. AAJ62207AE
Change view
Click to view available options
Quantity:
100 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ62207AE 100 mL
AAJ62207AK 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ62207AE Supplier Thermo Scientific Chemicals Supplier No. J62207AE

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

CAS 288-32-4
Concentration 0.5 M
pH 6.5
Physical Form Liquid
Quantity 100 mL
Molecular Formula C3H4N2
MDL Number MFCD00005183
InChI Key RAXXELZNTBOGNW-UHFFFAOYSA-N
SMILES N1C=CN=C1
IUPAC Name 1H-imidazole
Molecular Weight (g/mol) 68.08
Show More Show Less