missing translation for 'onlineSavingsMsg'
Learn More

Imidacloprid, SPEX CertiPrep™

Catalog No. S2247
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S2247 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S2247 Supplier SPEX CertiPrep Group LLC Supplier No. S2247

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

AnalyteCAS No.Concentration
Imidacloprid138261-41-31000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Chemical Identifiers

CAS 138261-41-3 , 67-64-1
Molecular Formula C9H10ClN5O2
Molecular Weight (g/mol) 255.662
InChI Key YWTYJOPNNQFBPC-UHFFFAOYSA-N
PubChem CID 86418
ChEBI CHEBI:5870
IUPAC Name N-[1-[(6-chloropyridin-3-yl)methyl]-4,5-dihydroimidazol-2-yl]nitramide
SMILES C1CN(C(=N1)N[N+](=O)[O-])CC2=CN=C(C=C2)Cl

Specifications

Packaging 2mL Amber Prescored Glass Ampule
Concentration or Composition Notes Certificate of Analysis reports target analytes to a ±2% accuracy and correct for raw material purity.
CAS Min % 0.1, 99.9
CAS Max % 0.1, 99.9
For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
Type Certified Reference Material
Solvent or Matrix Acetone
Quantity 1 mL
Chemical Name or Material Imidacloprid
Hazard Category Harmful if swallowed. Suspected of causing cancer.
DOTInformation Nonhazardous
Recommended Storage Ambient

Product intended for laboratory use only