missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Imidacloprid

CAS: 138261-41-3 | C9H10ClN5O2, C9H10ClN5O2 | 255.66 g/mol

$497.34 - $497.34

Chemical Identifiers

CAS 138261-41-3
Molecular Formula C9H10ClN5O2
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC466750010
View Documents
Thermo Scientific Chemicals
466750010
1 g
Each for $497.34
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

138261-41-3
C9H10ClN5O2

Specifications

138261-41-3
134.0°C to 146.0°C
255.66
[O-][N+](=O)NC1=NCCN1CC1=CC=C(Cl)N=C1
255.66
1 g
C9H10ClN5O2
YWTYJOPNNQFBPC-UHFFFAOYSA-N
2-chloro-5-{[2-(nitroamino)-4,5-dihydro-1H-imidazol-1-yl]methyl}pyridine
Imidacloprid
Videos
Safety and Handling

Safety and Handling

GHS H Statement Very toxic to aquatic life with long lasting effects.

Harmful if swallowed.

GHS P Statement IF SWALLOWED: rinse mouth.

Do NOT induce vomiting.

Call a POISON CENTER or doctor/physician if you feel unwell.

Wash face, hands and any exposed skin thoroughly after handling.

Avoid release to the environment.

Collect spillage.

Dispose of contents/container to an approved waste disposal plant.

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only