missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Image-iT™ Green Hypoxia Reagent
Image-iT Green Hypoxia Reagent is a novel, fixable fluorogenic compound for measuring hypoxia in live cells.
Supplier: Invitrogen™ I14833
Description
Image-iT Green Hypoxia Reagent is a novel, fixable fluorogenic compound for measuring hypoxia in live cells. It is non-fluorescent when live cells are in an environment with normal oxygen concentrations and becomes fluorescent when oxygen levels are decreased. Image-iT Green Hypoxia Reagent sustains its fluorescence when cells/tissue return to normal oxygen levels, allowing the cells/tissue to be fixed with minimal loss of fluorescent signal.Features of Image-iT Green Hypoxia Reagent include:
• Indicates hypoxia in live cells by fluorescing in low oxygen environments
• Fixable—sustains fluorescent signal when cells/tissue return to normal oxygen levels
• Easy to use—just add to cell culture media and image
Image-iT Green Hypoxia Reagent is a fluorogenic compound that is live cell permeable and becomes fluorescent in environments with low oxygen concentrations. These properties makes it a highly useful tool for detecting cells and tissue under hypoxic conditions. Image-iT Green Hypoxia Reagent is a very sensitive oxygen detector. Unlike pimonidazole adducts that respond only to very low oxygen levels, Image-iT Green Hypoxia Reagent begins to fluoresce when atmospheric oxygen levels are less than 5%. It responds quickly to such environments, and the fluorescence is sustained after the oxygen levels return to normal. These properties make Image-iT Green Hypoxia Reagent an ideal tool for detecting hypoxic conditions around tumors, 3D cultures, spheroids, neurons, etc.
Order Info
- Store at -5 to -30°C. Protect from light.
- Stable for at least 6 months after receiving.
Specifications
| Store at -5 to -30°C. Protect from light. |
|
| Image-iT | |
| Hypoxia Reagent |
| Reagent | |
| 5 Vials | |
| Approved for shipment on Wet or Dry Ice |
For Research Use Only. Not for use in diagnostic procedures.