missing translation for 'onlineSavingsMsg'
Learn More

Cytiva IMAC Sepharose™ High Performance Media

Catalog No. 45000166
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45000166 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45000166 Supplier Cytiva Supplier No. 17092007

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For optimizing purification of histidine-tagged proteins using Immobilized Metal Affinity Chromatography (IMAC) when Ni2+ is not the best choice of metal ion

  • 34μm beads of highly cross-linked 6% agarose, to which a chelating group has been covalently coupled
  • Charge with transition metal of choice for optimized selectivity
  • Small bead size for high performance purification
  • High binding capacity: ≥40mg histidine-tagged protein/mL medium when charged with Ni2+
  • Allows more sample to be applied; a more concentrated target protein is obtained more at a lower cost/mg and in less time
  • Stable in pH range 3-12 (long term), and in all commonly used buffers

Avoid use with chelating agents, e.g., EDTA, EGTA, or citrate.

Specifications

Flow Rate <300 cm/hr.
pH Range 3 to 12
Particle Size 34 μm
Product Type IMAC Sepharose HP
Content And Storage 4°C to 30°C
Quantity 100 mL
For Use With (Application) Purification of Polyhistidine tagged Proteins