missing translation for 'onlineSavingsMsg'
Learn More

Cytiva triplePrep Kit

Catalog No. 45001810
Change view
Click to view available options
No. of Reactions:
50 Preps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
45001810 50 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45001810 Supplier Cytiva Supplier No. 28942544
NLSU: Get 1 Consumable Product When You Buy 1 Obtenez 1 produit consommable à l’achat d’un produit
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

Protocol and a supply of components sufficient for 50 preps.

Isolate gDNA, total RNA, and total denatured proteins from undivided animal tissues and mammalian cell samples in less than 1 hour

  • High-yield process allows extraction/isolation from single limited-size samples from biopsies, archived tissues, and tumors
  • Isolated products suitable for downstream applications such as PCR, restriction digestion, sequencing, array CGH, RT-qPCR, gene expression microarray, SDS-PAGE, Western blotting, 2-D DIGE, and LCMS
  • Flexible workflow allows easy isolation of any two or all three analytes
  • Color-coded caps and bottles match to protocol steps to minimize the chance for error

Specifications

Description The illustra triplePrep Kit is designed for rapid, simultaneous extraction and isolation of high quality genomic DNA (gDNA), total RNA, and total denatured proteins from animal tissues and mammalian cells.
Format Kit
Isolation Technology Silica Spin Column
For Use With (Application) Extraction and isolation of high quality genomic DNA, total RNA and total denatured Proteins from animal tissues and mammalian cells
Includes Protocol and a supply of components sufficient for 50 preps
No. of Reactions 50 Preps
Product Type illustra triplePrep kit