Learn More
Cytiva illustra™ Ready-To-Go GenomiPhi V3 DNA Amplification Kit
Used for various applications like genotyping (SNP, STR, array CGH), cloning, sequencing, and DNA archival. Cytiva illustra™ Ready-To-Go GenomiPhi V3 DNA Amplification Kit yeilds high-quality DNA.
Supplier:  Cytiva 25660196
Catalog No. 45002384
Only null left
Description
Description
  • Representative amplification of the whole genome 12 to 20μg yield from nanogram amounts of DNA sample.
  • Preformulated, predispensed, single-dose, lyophilized cakes for rapid reaction set-up and greater reproducibility
  • Quick and simple automation-friendly protocol; no thermal cycler required
  • Sample Weight: 10ng of genomic DNA; or non-purified cell lysates
  • Product Size Avg: >10kb
  • Shipping and storage at room temperature provides easy access and reduced carbon footprint
Order Info
Custom options of the product, including larger pack sizes, are available. Please contact your local sales representative or Technical Support.
Specifications
Specifications
Ready-To-Go GenomiPhi V3 cakes, 1 × 96-well plate; 2x Denaturation buffer, 1 × 1.2ml; Control DNA (Lambda), 500ng; 12 Strips of 8 domed caps. Store the kit at ambient temperature. Once opened, completely reseal the pouch containing the Ready-To-Go GenomiPhi V3 DNA amplification kit cakes. To ensure maximum shelf life and optimal performance, storage in a desiccator is recommended. Store reconstituted Control DNA (Lambda) at −20°C. Once opened, store 2× denaturation buffer at 2 to 8°C
96 reactions
Genotyping
Product Suggestions
Product Suggestions