missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva RAPD Analysis Beads
For performing DNA-profiling experiments using the randomly amplified polymorphic DNA (RAPD) technique
$1,230.34 - $2,105.43
Specifications
| Format | Tube |
|---|---|
| pH | 8.3 |
| For Use With (Application) | Standard PCR |
| Volume | 500 μL |
Description
- Preformulated, single-dose, ambient-temperature-stable reaction beads ensure greater reproducibility between reactions, minimize pipetting steps and reduce the potential for pipetting errors and contamination
- RAPD reactions are simple to perform—simply add genomic DNA solution and primer to a tube of RAPD Analysis Beads to a final reaction volume of 25 μL and cycle the reaction
- Each lot of RAPD Analysis Beads is function-tested to ensure its ability to generate a differential banding pattern between the two control E. coli strains using RAPD Analysis Primer 2 included with the RAPD beads
- RAPD reactions are preoptimized for use with a wide variety of organisms
- Beads are provided in thin-walled 0.5 mL tubes compatible with most thermal cyclers
Specifications
| Tube | |
| 8.3 | |
| Standard PCR | |
| 500 μL |