missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva NAP™ Columns, NAP-25
Use for rapid and efficient purification of DNA and oligonucleotides (>10-mer) in less than 15 minutes by gravity flow. Cytiva Life Sciences™ illustra™ NAP™ Columns are disposable columns prepacked with Sephadex™ G-25 DNA Grade
$425.00 - $907.00
Specifications
| For Use With (Application) | Rapid and efficient purification of DNA and Oligonucleotides |
|---|---|
| Includes | Complete step-by-step protocol identifying the fixed volumes needed for equilibration, washing and sample elution |
| Product Type | NAP-25 |
Includes: Complete step-by-step protocol identifying the fixed volumes needed for equilibration, washing, and sample elution
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
45-000-149
|
Cytiva
17085201 |
20 Pack |
Pack of 20 Each for $425.00
|
|
|||||
|
45-000-150
|
Cytiva
17085202 |
50 Pack |
Pack of 50 Each for $907.00
|
|
|||||
Description
- Disposable columns prepacked with Sephadex™ G-25 DNA Grade Gel require only gravity to run
- Available in three sizes depending on sample volume: 0.5mL (NAP-5), 1mL (NAP-10) or 2.5mL (NAP-25)
- Useful for small-scale purification, desalting, and buffer exchange
Order Info
Sephadex G-25 DNA grade gel comes in a column buffer consisting of distilled water with 0.15% Kathon™ CG/ICP Biocide.
Specifications
| Rapid and efficient purification of DNA and Oligonucleotides | |
| Complete step-by-step protocol identifying the fixed volumes needed for equilibration, washing and sample elution | |
| NAP-25 |