Learn More
Cytiva MicroSpin™ S-200 HR Columns
For rapid purification of labeled single-stranded or double-stranded DNA fragments ≥50 bases in length in a volume of 25 to 100μL using spin-column chromatography
Supplier: Cytiva 27512001
Description
- Prepacked with Sephacryl™ S-200 HR medium pre-equilibrated in TE buffer
- Percent sample recovery: >80%
- Tested in nickase, single- and double-stranded exonuclease and RNase assays
Specifications
| Spin Column | |
| Rapid purification of labeled single-stranded or double-stranded DNA fragments <50 bases in length in a volume of 25 to 100μL using spin-column chromatography | |
| Column | |
| Each |