missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Microspin G-50 Columns

Catalog No. 45001399
Change view
Click to view available options
No. of Reactions:
250 Preps
50 Preps
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
45001399 250 Preps
45001398 50 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 45001399 Supplier Cytiva Supplier No. 27533002
NLSU: Get 1 Consumable Product When You Buy 1 Obtenez 1 produit consommable à l’achat d’un produit
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Designed to allow the rapid purification of DNA for use in a wide range of applications using typical microfuge equipment. Cytiva illustra™ Microspin G-50 Columns will not remove or denature enzyme.

  • Optimal sample volumes in the range of 12-50μ
  • For rapid purification of labeled DNA (>20 bases) from smaller molecules in solution
  • Columns are prepacked with Sephadex G-50 Grade equilibrated in TE buffer with 0.05% Kathon CG/ICP Biocide
  • illustra AutoSeq™ and illustra ProbeQuant™ G-50 Micro Columns are specifically designed for dye terminator removal and labelled probe purification respectively and offer optimized performance in those applications
  • Can also be used for buffer exchange/desalting of enzymatic reactions (10 to 100 μL) following heat denaturation and phenol extraction
  • Product recovery will be at least 10% lower than that from ProbeQuant G-50 Micro Columns.
  • For rapid purification of labeled DNA (>20 bases) from unincorporated labeled nucleotides in a volume of 25 to 50 μl using spin-column chromatography

 

Desalting, buffer exchange; Removal of dye terminators from cycle sequencing reactions; Removal of labeled nucleotides from DNA labeling reactions

Order Info

Conveniently prepacked with Sephadex G-50 DNA Grade pre-equilibrated in TE buffer with 0.05% Kathon™ CG/ICP Biocide and collection tubes

Specifications

Format Column
Isolation Technology Spin Column
For Use With (Application) Desalting, Buffer exchange, removal of dye terminators from cycle sequencing reactions, removal of labeled, Nucleotides in 25 to 50μL from DNA labeling reactions
Includes Sephadex™ G-50 DNA grade
No. of Reactions 250 Preps
Product Type Column