missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva GFX™ PCR DNA and Gel Band Purification Kit
Purify and concentrate PCR products or DNA fragments ranging in size from 50 bp to 10 kb. Cytiva Life Sciences™ illustra™ GFX™ PCR DNA and Gel Band Purification Kit can be used to purify DNA from reaction volumes up to 100μL or agarose gel slices of up to 900mg.
$333.69 - $820.00
Specifications
| Description | Designed for the rapid purification and concentration of PCR products or DNA fragments ranging in size from 50 bp to 10 kb. This kit can be used to purify DNA from reaction volumes up to 100 μl or agarose gel slices up to 900 mg. |
|---|---|
| Sample Type | Agarose Gel Slices, DNA Solutions, Enzyme Reactions, PCR Mixtures |
| Formulation | Tris HCl and sterile Water |
| For Use With (Application) | Isolation and concentration of DNA fragments (50bp to 10kb) from PCR mixtures, DNA-containing Agarose Gel bands, Enzyme-based DNA modifications and restriction digestions |
| Includes | GFX™ columns, collection tubes, color-coded bottles of capture Buffer, Wash Buffer, two Elution Buffers |
Includes: GFX columns; Collection tubes; Color-coded bottles of capture buffer; Wash buffer; two elution buffers: Tris-HCl and sterile water
| Catalog Number | Mfr. No. | No. of Reactions | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | No. of Reactions | Price | Quantity | |||||
|
45001489
|
Cytiva
28903470 |
100 Preps |
N/A
|
|
|||||
|
45-001-490
|
Cytiva
28903471 |
250 Preps |
N/A
|
|
|||||
Description
Typical recoveries range from 60 to 80% for DNA fragments from agarose gel to as high as 95% for PCR products from solution.
- Rapid and easy-to-use purification method with less than 10 minutes hands-on time
- Flexible 10 to 50μL elution volume for different DNA concentration needs
- Improved column design with increased maximum volume holding of up to 800μL liquid volume (950μL without a cap)
- DNA of exceptional purity (to 99.5%) is ready for direct use in sequencing, PCR, labeling, restriction enzyme digestion, and cloning
- Visual color indicator added to the capture buffer to ensure optimal pH for maximum DNA binding and recovery
- Kit combines a versatile chaotropic buffer with a glass fiber matrix supported in a spin-column for the purification of DNA from both solution and agarose gel
Specifications
| Designed for the rapid purification and concentration of PCR products or DNA fragments ranging in size from 50 bp to 10 kb. This kit can be used to purify DNA from reaction volumes up to 100 μl or agarose gel slices up to 900 mg. | |
| Tris HCl and sterile Water | |
| GFX™ columns, collection tubes, color-coded bottles of capture Buffer, Wash Buffer, two Elution Buffers | |
| 50 bp-10 kb |
| Agarose Gel Slices, DNA Solutions, Enzyme Reactions, PCR Mixtures | |
| Isolation and concentration of DNA fragments (50bp to 10kb) from PCR mixtures, DNA-containing Agarose Gel bands, Enzyme-based DNA modifications and restriction digestions | |
| illustra GFX PCR DNA and Gel Band Purification Kit | |
| PCR 10 μl elution volume-65%, PCR 50 μl elution volume-82%, 300 mg agarose 10 μl elution volume-57%, 300 mg agarose 50 μl elution volume-91% |