missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva GenomiPhi HY DNA Amplification Kit
Highly efficient and representative whole-genome amplification yielding 40 to 50μg from nanogram amounts of DNA sample
$1102.00 - $3439.00
Specifications
| Content And Storage | Store the kit at -70°C.The enzyme mix must be stored at -70°C; all other components may be stored at -20°C.Thaw components on ice and maintain at 0°C to 4°C during handling. |
|---|---|
| Description | Illustra GenomiPhi HY DNA Amplification Kit offers highly efficient and representative whole-genome amplification with 40 to 50 μg yield from nanogram amounts of DNA sample. |
| For Use With (Application) | Genomic DNA Amplification |
| Product Type | illustra GenomiPhi HY DNA Amplification Kit |
| Size | > 10 kb |
| Catalog Number | Mfr. No. | No. of Reactions | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | No. of Reactions | Price | Quantity | |||||
|
45-001-218
|
Cytiva
25660020 |
100 reactions |
Each for $3,439.00
|
|
|||||
|
45-001-219
|
Cytiva
25660022 |
25 reactions |
Each for $1,102.00
|
|
|||||
Description
- Starting material can be purified DNA or nonpurified cell lysates
- Simple protocol provides typical DNA of 40 to 50μg, average lengths >10kb
- Less hands-on time compared to traditional isolation methods
- Representative amplification of the whole genome
- Automation-friendly protocol with great reproducibility
- High-quality DNA for array CGH, high-throughput genotyping, DNA archival, PCR, restriction enzyme digestion, hybridization, and cloning
- Contains all of the components necessary for genomic DNA preparation by isothermal strand displacement amplification
Specifications
| Store the kit at -70°C.The enzyme mix must be stored at -70°C; all other components may be stored at -20°C.Thaw components on ice and maintain at 0°C to 4°C during handling. | |
| Genomic DNA Amplification | |
| > 10 kb |
| Illustra GenomiPhi HY DNA Amplification Kit offers highly efficient and representative whole-genome amplification with 40 to 50 μg yield from nanogram amounts of DNA sample. | |
| illustra GenomiPhi HY DNA Amplification Kit | |
| 4 h |