missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Exonuclease I

Catalog No. 45500137
Change view
Click to view available options
Quantity:
2500 U
5000 U
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45500137 2500 U
45001407 5000 U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 45500137 Supplier Cytiva Supplier No. E70073Z
NLSU: Get 1 Consumable Product When You Buy 1 Obtenez 1 produit consommable à l’achat d’un produit
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Catalyzes sequential removal of nucleotides 3' end of single-stranded DNA molecule

  • Monomeric and bacterial
  • Molecular weight: 54.5kd
  • Expressed in E.Coli
  • Inactivated by heating at 80°C for 15 minutes
  • Active in PCR buffers
  • Activity: 10U/μL
  • Free of contaminating endonucleases, double-stranded exonucleases and ribonucleases
  • >95% pure by SDS-PAGE

Specifications

Purity >95% by SDS-PAGE
Source Bacteria
Quantity 2500 U
Product Type Exonuclease I
Molecular Weight (g/mol) 54.5kd