missing translation for 'onlineSavingsMsg'
Learn More

IL17D Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™

Rabbit Polyclonal Antibody

$337.00 - $432.00

Specifications

Antigen IL17D
Concentration 0.1-1.0 mg/mL
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Biotin
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
500P88BT25 DFS Item
View Documents
PeproTech
500P88BT25UG
25 μg
Each for $337.00
Only null left
 
500P88BT50 DFS Item
View Documents
PeproTech
500P88BT50UG
50 μg
Each for $432.00
Only null left
 
50-271-8437
View Documents
PeproTech
500P88BT500UG
500 μg N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

AA Sequence of recombinant protein: APRAGRRPAR PRGCADRPEE LLEQLYGRLA AGVLSAFHHT LQLGPREQAR NASCPAGGRP ADRRFRPPTN LRSVSPWAYR ISYDPARYPR YLPEAYCLCR GCLTGLFGEE DVRFRSAPVY MPTVVLRRTP ACAGGRSVYT EAYVTIPVGC TCVPEPEKDA DSINSSIDKQ GAKLLLGPND APAGP. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human IL-17D. Anti-Human IL-17D-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect Human IL-17D by sandwich ELISA (using 100 mL/well) a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti- Human IL-17D (500-P88) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Human IL-17D. Western Blot: To detect Human IL-17D by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 mg/mL. when used in conjunction with compatible development reagents the detection limit for Recombinant Human IL-17D is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. 500-P88BT-1MG will be provided as 2 x 500 μg

The protein encoded by this gene is a cytokine that shares the sequence similarity with IL17. The treatment of endothelial cells with this cytokine has been shown to stimulate the production of other cytokines including IL6, IL8 and CSF2/ GM-CSF. The increased expression of IL8 induced by this cytokine was found to be NF-kappa B-dependent.
Specifications

Specifications

IL17D
ELISA, Western Blot
Biotin
Rabbit
Human
Q8TAD2
53342
E.coli-derived Recombinant Human IL-17D.
Antigen affinity chromatography
IL17D
0.1-1.0 mg/mL
Polyclonal
Lyophilized
RUO
PBS with no preservative
AI462269; FLJ30846; IL17D; IL-17D; IL-22; IL-27; Il27a; IL-3R; Interleukin; interleukin 17D; interleukin 27A; interleukin-17D; Interleukin-27; UNQ3096/PRO21175
IL17D
Primary
-20°C
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'