Learn More
PeproTech IL17D Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: PeproTech 500P88BT500UG
Description
AA Sequence of recombinant protein: APRAGRRPAR PRGCADRPEE LLEQLYGRLA AGVLSAFHHT LQLGPREQAR NASCPAGGRP ADRRFRPPTN LRSVSPWAYR ISYDPARYPR YLPEAYCLCR GCLTGLFGEE DVRFRSAPVY MPTVVLRRTP ACAGGRSVYT EAYVTIPVGC TCVPEPEKDA DSINSSIDKQ GAKLLLGPND APAGP. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human IL-17D. Anti-Human IL-17D-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect Human IL-17D by sandwich ELISA (using 100 mL/well) a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti- Human IL-17D (500-P88) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Human IL-17D. Western Blot: To detect Human IL-17D by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 mg/mL. when used in conjunction with compatible development reagents the detection limit for Recombinant Human IL-17D is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. 500-P88BT-1MG will be provided as 2 x 500 μg
The protein encoded by this gene is a cytokine that shares the sequence similarity with IL17. The treatment of endothelial cells with this cytokine has been shown to stimulate the production of other cytokines including IL6, IL8 and CSF2/ GM-CSF. The increased expression of IL8 induced by this cytokine was found to be NF-kappa B-dependent.Specifications
| IL17D | |
| Polyclonal | |
| Biotin | |
| IL17D | |
| AI462269; FLJ30846; IL17D; IL-17D; IL-22; IL-27; Il27a; IL-3R; Interleukin; interleukin 17D; interleukin 27A; interleukin-17D; Interleukin-27; UNQ3096/PRO21175 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 53342 | |
| -20°C | |
| Lyophilized |
| ELISA, Western Blot | |
| 0.1-1.0 mg/mL | |
| PBS with no preservative | |
| Q8TAD2 | |
| IL17D | |
| E.coli-derived Recombinant Human IL-17D. | |
| 500 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |