missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Mouse IL-2 Uncoated ELISA Kit

ELISA

$1060.00 - $1444.00

Specifications

Accession Number P04351
Assay Range 2 to 200 pg/mL
Assay Sensitivity 2 pg/mL
Conjugate HRP
Product Type ELISA
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5017190
View Documents
Invitrogen™
88702477
20 x 96 Tests
Each for $1,444.00
Only null left
 
5017192
View Documents
Invitrogen™
88702488
10 x 96 Tests
Each for $1,060.00
Only null left
 
Description

Description

The Mouse Interleukin-2 (IL-2) Uncoated ELISA Kit contains pre-matched antibody pairs, and reagents for performing quantitative enzyme linked immunosorbent assays (ELISA) to detect and quantify protein levels of mouse IL-2. Wash Buffer and Stop Solution are needed to complete the ELISA reaction and are sold separately. Principle of the method ELISAs are designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody is coated to the bottom of the wells of a microplate, which is an overnight process. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. A sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen.

Interleukin 2 (IL-2) is an immuno-modulatory cytokine that is important for the proliferation of activated T cells, differentiation of B cells, natural killer cells, monocytes and macrophages. IL-2 signals through the IL-2 receptor (IL-2R), a heterotrimeric protein complex whose gamma chain is also shared by interleukin 4 (IL-4) and interleukin 7 (IL-7). The expression of the IL-2 gene in mature thymocytes is monoallelic, which represents an unusual regulatory mode for controlling the precise expression of a single gene. The targeted disruption of a gene similar to IL-2 in mice leads to an ulcerative colitis-like disease that suggests an essential role of this gene in the immune response to antigenic stimuli.
Specifications

Specifications

P04351
2 pg/mL
ELISA
Mouse
Colorimetric Microplate Reader
DIF-alpha,TNFA,TNFSF2,TNLG1F,TNF
HRP
Serum 100 μL, Supernatant 100 μL
Interleukin-2
1 hrs 45 mins
2 to 200 pg/mL
HRP
Serum, Supernatant
ELISA
16183
Capture Antibody: Pre-titrated, 96 Well Plate: High affinity 96-well plate (included with product Cat. Nos. ending in suffixes -22, -76, -86), Detection Antibody: Pre-titrated, Detection Antibody: Pre-titrated, Standard: Recombinant protein for generating standard curve and calibrating samples, Standard: Recombinant protein for generating standard curve and calibrating samples, 5X ELISA/ELISPOT Diluent: Buffer for blocking and diluting the Detection Antibody and Enzyme, 5X ELISA/ELISPOT Diluent: Buffer for blocking and diluting the Detection Antibody and Enzyme, Substrate: 1X TMB Solution, Substrate: 1X TMB Solution, Certificate of Analysis: Lot-specific instructions for dilution of antibodies and standards
RUO
2°C to 8°C
IL-2
24 hrs 30 mins
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'