missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Igepal Ca - 630 Non-Ionic Detergent MP Biomedicals
$65.84 - $214.82
Chemical Identifiers
| CAS | 9036-19-5 |
|---|---|
| Molecular Formula | (C2H4O)x·C14H22O |
| Synonym | Polyethylene glycol octylphenol ether |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
ICN19859650
|
N/A | 50 mL |
Each for $65.84
|
|
|||||
|
ICN19859680
|
N/A | 100 mL |
Each for $104.86
|
|
|||||
|
ICN19859690
|
N/A | 500 mL |
Each for $214.82
|
|
|||||
Chemical Identifiers
| 9036-19-5 | |
| (C2H4O)x·C14H22O | |
| Polyethylene glycol octylphenol ether |
Specifications
| 9036-19-5 | |
| (C2H4O)x·C14H22O | |
| Igepal Ca - 630, Non-Ionic Detergent |
| 1.06 g/mL | |
| Polyethylene glycol octylphenol ether |