missing translation for 'onlineSavingsMsg'
Learn More

Promega IdeZ Protease, 5,000 units

Catalog No. PRV8341
Encompass
Change view
Click to view available options
Quantity:
5000 U
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRV8341 5000 U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRV8341 Supplier Promega Supplier No. V8341

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

IdeS Protease is an immunoglobulin-degrading enzyme from Streptococcus pyogenes (IdeS). IdeZ Protease is an immunoglobulin-degrading enzyme from Streptococcus equi subspecies zooepidemicus.

  • Antibody Fragmentation and Characterization, Simplified
  • Highly specific IgG cleavage
  • Rapid generation of highly pure fragments
  • Frozen protease is supplied at a concentration of 50u/μl

Specifications

Content And Storage <-10°C
Quantity 5000 U