missing translation for 'onlineSavingsMsg'
Learn More

ITV Ice Makers Ice Machine with Bin

Cube-style ice maker with ice bin.

$7,737.55 - $10,614.17

Specifications

Product Type Cube Ice Maker
Voltage 115 V
View More Specs

Products 4
Catalog Number Mfr. No. Yield Price Quantity & Availability  
Catalog Number Mfr. No. Yield Price Quantity & Availability  
11675155
View Documents Promotion Details
ITV Ice Makers
SPIKA NG 130
134 lb. (60 kg)
Each for $7,737.55
Only null left
 
11675156
View Documents Promotion Details
ITV Ice Makers
SPIKA NG 160
159 lb. (72 kg)
Each for $8,839.45
Only null left
 
11675153
View Documents Promotion Details
ITV Ice Makers
SPIKA NG 230
223 lb. (205 kg)
Each for $9,510.70
Only null left
 
11675154
View Documents Promotion Details
ITV Ice Makers
SPIKA NG 360
353 lb. (160 kg) N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

  • SPIKA Ice Maker
  • Self-contained
  • Half or full Classic American ice cube-style
  • Electro mechanic timer
  • Dual switch
  • 304 stainless steel construction
  • Vertical plate evaporator
  • Legs adjustable from 4 to 6 in. (not included on dimensions)
  • cETLus, NSF
Specifications

Specifications

Cube Ice Maker
115 V
Videos
SDS
Documents

Documents