missing translation for 'onlineSavingsMsg'
Learn More

ITV Ice Makers Ice Machine with Bin

Cube-style ice maker with ice bin.

Supplier:  ITV Ice Makers SPIKA NG 360

 View more versions of this product

Catalog No. 11675154

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

  • SPIKA Ice Maker
  • Self-contained
  • Half or full Classic American ice cube-style
  • Electro mechanic timer
  • Dual switch
  • 304 stainless steel construction
  • Vertical plate evaporator
  • Legs adjustable from 4 to 6 in. (not included on dimensions)
  • cETLus, NSF
Specifications

Chemical Identifiers

Cube Ice Maker
115 V

Specifications

99 lb.
Cube Ice Maker
353 lb. (160 kg)
45 kg
115 V
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions