missing translation for 'onlineSavingsMsg'
Learn More

Ibuprofen sodium salt, 98% (GC), MilliporeSigma™ Supelco™

Ibuprofen belongs to the class of non-steroidal anti-inflammatory drugs, widely used for the treatment of mild to moderate pain and several forms of arthritis.

$356.20 - $1318.71

Specifications

CAS 31121-93-4
Physical Form Neat
Linear Formula C13H17O2 Na
MDL Number MFCD00133414
Synonym alpha-Methyl-4-(isobutyl)phenylacetic acid
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
111013894
View Documents
Merck Emd Millipore
I1892100G
100 g
Each for $356.20
Only null left
 
111013893
View Documents
Merck Emd Millipore
I1892500G
500 g
Each for $1,318.71
Only null left
 
Description

Description

Featured Industries: Pharmaceutical (small molecule)
Specifications

Specifications

31121-93-4
C13H17O2 Na
alpha-Methyl-4-(isobutyl)phenylacetic acid
Analytical Standard
≥98% (GC)
Neat
MFCD00133414
Soluble in water
228.26
Analytical Standard
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'