missing translation for 'onlineSavingsMsg'
Learn More

Ibuprofen sodium salt, 98% (GC), MilliporeSigma™ Supelco™

Ibuprofen belongs to the class of non-steroidal anti-inflammatory drugs, widely used for the treatment of mild to moderate pain and several forms of arthritis.

Supplier:  Merck Emd Millipore I1892500G

 View more versions of this product

Catalog No. 111013893


Only null left
Explore available promotions

Description

Description

Featured Industries: Pharmaceutical (small molecule)
Specifications

Specifications

31121-93-4
C13H17O2 Na
alpha-Methyl-4-(isobutyl)phenylacetic acid
Analytical Standard
228.26
Analytical Standard
Neat
MFCD00133414
Soluble in water
500 g
≥98% (GC)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions