missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Power SYBR™ Green RNA-to-CT1-Step Kit

The novel formulation reduces false positive results caused by primer dimers, significantly improves data accuracy, and provides reliable detection of low abundance targets.

Supplier:  Applied Biosystems™ 4389986

Encompass

 View more versions of this product

Catalog No. 4389986


Get Up to $3,000 of qPCR Products with Purchase
View all promotions
Only null left

Description

Description

The novel formulation reduces false positive results caused by primer dimers, significantly improves data accuracy, and provides reliable detection of low abundance targets.

Get sensitivity and specificity in an easy-to-use one-step qRT-PCR reaction. The Power SYBR™ Green RNA-to-CT1-Step kit has been formulated to ensure maximum sensitivity and reliability. The novel formulation reduces false positive results caused by primer dimers, significantly improves data accuracy, and provides reliable detection of low abundance targets. It includes:

  • AmpliTaq™ Gold DNA Polymerase UP for hot start qPCR, minimizing non-specific product formation
  • ArrayScript™ UP Reverse Transcriptase, an engineered RT that produces high yields of full-length cDNA
  • An additive that reduces primer dimer formation, reducing non-specific products that can cause false positive signals
  • RNase inhibitor, to prevent degradation of RNA templates
  • A proprietary ROX™ dye that serves as a passive internal reference to normalize non-PCR-related fluorescence fluctuations for superb precision on Applied Biosystems™ real-time PCR instruments.

Order Info

Shipping Conditions: Wet Ice

Specifications

Chemical Identifiers

SYBR
AmpliTaq Gold DNA Polymerase
7500 Fast System, 7500 System, 7900HT System, QuantStudio™ 12k Flex, QuantStudio™ 3, QuantStudio™ 5, QuantStudio™ 6 Flex, QuantStudio™ 7, StepOne™, StepOnePlus™, ViiA™ 7 System
ROX (Pre-mixed)
UP (Ultra Pure)
Standard
Gene Expression
1-step RT-qPCR
Standard
SYBR Green
RNA-to-CT, SYBR
ArrayScript™ UP
Wet Ice
RNA

Specifications

Sufficient for 200 reactions at 20 μL reaction volume and includes:
• 1 x 5 mL tube of 2X master mix containing SYBR™Green 1 Dye, AmpliTaq Gold™ DNA Polymerase UP, dNTPs, Passive Reference 1,and optimized buffer components. Store at 2-8°C after opening.
• 1 x 80 μL tube of 125X RT enzyme mix containing ArrayScript™ UP Reverse Transcriptase, RNase Inhibitor. Store at -15 to -30°C.

Guaranteed minimum shelf life is 60 days (exact expiry date printed on product and CofA).
1-step RT-qPCR
Standard
SYBR Green
RNA-to-CT, SYBR
UP (Ultra Pure)
ArrayScript™ UP
Wet Ice
Gene Expression
RNA
SYBR
AmpliTaq Gold DNA Polymerase
7500 Fast System, 7500 System, 7900HT System, QuantStudio™ 12k Flex, QuantStudio™ 3, QuantStudio™ 5, QuantStudio™ 6 Flex, QuantStudio™ 7, StepOne™, StepOnePlus™, ViiA™ 7 System
ROX (Pre-mixed)
One-Step qRT-PCR Kit
200 Reactions
Standard
200 Reactions at 20 μL
200 Reactions
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.